Orders LinkContactUsLinkComponent
Round Bottom Flasks
Round Bottom Flasks
250 ml, 24/29, Round
Catalog # 47800-146
Supplier:   Heidolph NA, LLC
Supplier #:  015890720
Round Bottom Flasks
Catalog # 47800-146
Supplier:  Heidolph NA, LLC
Round Bottom Flasks
250 ml, 24/29, Round
Price
$342.62
Log in for availability
Pack of 2
Minimum order quantity is 1
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
  • [ST] joint no.:
    24/29
  • Capacity:
    250 ml
  • Bottom style:
    Round