$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
KF STABLE TITRANT 1603 BOTTLE 2MG
34126-302
:
GFS Chemicals
KF STABLE TITRANT 1603 BOTTLE 2MG
34126-302
:
GFS Chemicals
:
99651
ghs_1603.pdf
VWR GHS Compliance Letter (US)