J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
Product Details & Documents
This BAKER ANALYZED® Reagent is a phosphate buffer solution for laboratory, research, and/or manufacturing uses.
- Contains no presevative
Each lot is compared to NIST Standard Reference Buffer.
Documents
Certificates
Search by Lot Number5656 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT5656-3
Product specification
Specifications
Specifications
- Pack type:Cubetainer®
- Color:Yellow
- Size:4 L
- Reference temperature:25 °C
- pH:7.00
- pH accuracy:± 0.04
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



