Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Polysorbate 20

Polysorbate 20

Bottle, 100 ml
Catalog # AAAL15029-AE
Supplier #:  L15029-AE
Polysorbate 20
Catalog # AAAL15029-AE
CAS Number:  9005-64-5
Polysorbate 20

Bottle, 100 ml

CAS Number:  9005-64-5
Price
$45.69
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Used for solubilizing membrane proteins during isolation and purification

CAS No.: 9005-64-5 Molecular Formula: C58H114O26 Flash Point: 110°(230°F) Refractive Index: 1.4700 Boiling Point: 100° MDL No.: MFCD00165986

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Density:1.105
  • Flash Pt:>110 °C(230 °F)
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.