Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3-Nonen-2-one 96%

3-Nonen-2-one 96%

250g
Catalog # AAAA19084-30
Supplier #:  A19084-30
3-Nonen-2-one 96%
Catalog # AAAA19084-30
CAS Number:  14309-57-0
3-Nonen-2-one 96%
3-Nonen-2-one 96%

250g

CAS Number:  14309-57-0
Price
$295.21
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 96% CAS No.: 14309-57-0 Molecular Formula: C9H16O Formula Weight: 140.23 Flash Point: 81°(177°F) Refractive Index: 1.4490 Boiling Point: 85°/12mm Beilstein Reference: 1743558 MDL No.: MFCD00010241

Synonyms:  Non-3-en-2-one
  • CAS Number:14309-57-0
  • Formula:C₉H₁₆O
  • MDL Number:MFCD00010241
  • Molecular Weight:140.23 g/mol
  • Boiling Pt:85 °C (12 mmHg)
  • Boiling Pt:85 °C/12 mm
  • Density:0.848
  • Flash Pt:81 °C (177 °F)
  • Flash Pt:81 °C(177 °F)
  • Storage Temperature:Ambient
  • EINECS:238-248-9
Specifications

Specifications

  • Size:
    250g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.