Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Acrylamide 98+%

bottle, 100 g
Catalog # AAAA17157-22
Toxic
Health
Acrylamide 98+%
Catalog # AAAA17157-22
Supplier:  
CAS Number:  79-06-1
Acrylamide 98+%
Acrylamide 98+%

bottle, 100 g

CAS Number:  79-06-1
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ACRYLAMIDE 98% 100G Catalog #BT215245-100G

 

ACRYLAMIDE 100GRAMS Catalog #101643-092

 

ACRYLAMIDE 25GM Catalog #TCA1132-025G

 

See more alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98+% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Odor: Odorless Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032

Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Merck Index:14,129
  • Boiling Pt:125 °C/25 mm
  • Melting Pt:82…85 °C
  • Density:1.322
  • Flash Pt:138 °C (280 °F)
  • Storage Temperature:Ambient
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.