Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dimethyl glyoxime 99%

Dimethyl glyoxime 99%

bottle, 500 g
Catalog # AAAA12810-36
Flammable
Toxic
Supplier #:  A12810-36
Dimethyl glyoxime 99%
Catalog # AAAA12810-36
CAS Number:  95-45-4
Dimethyl glyoxime 99%
Dimethyl glyoxime 99%

bottle, 500 g

CAS Number:  95-45-4
Price
$154.51
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 95-45-4 Molecular Formula: C4H8N2O2 Formula Weight: 116.12 Melting Point: ca 240° dec. Beilstein Reference: 506731 Merck Reference: 14,3246 MDL No.: MFCD00002117

Synonyms:  Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
  • CAS Number:95-45-4
  • Formula:CH₃C(=NOH)C(=NOH)CH₃
  • MDL Number:MFCD00002117
  • Molecular Weight:116.12 g/mol
  • Merck Index:14,3246
  • Boiling Pt:53 °C (8 -rr)
  • Melting Pt:∼ 240 °C (dec.)
  • Density:1.37 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-420-1
  • UN:2926
  • ADR:4.1,III

Documents

Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.