Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ethanolamine 98+%

bottle, 250 g
Catalog # AAAA11697-30
Corrosive
Irritant
Ethanolamine 98+%
Catalog # AAAA11697-30
Supplier:  
CAS Number:  141-43-5
Ethanolamine 98+%
Ethanolamine 98+%

bottle, 250 g

CAS Number:  141-43-5
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ETHNLAMINE 500GM Catalog #TCA0297-500G

 

ETHANOLAMINE FOR SYNTH (100ML) Catalog #EM8.00849.0100

 

ETHANOLAMINE 500G Catalog #BT129135-500G

 

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98+% CAS No.: 141-43-5 Molecular Formula: C2H7NO Formula Weight: 61.08 Flash Point: 93°(199°F) Refractive Index: 1.4540 Odor: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183

Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:14,3727
  • Boiling Pt:169…170 °C
  • Melting Pt:10 °C
  • Density:1.012
  • Flash Pt:93 °C (199 °F)
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.