Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Hydrocortisone 98%

Bottle, 1 g
Catalog # AAA16292-03
Health
 :  A16292-03
Hydrocortisone 98%
Catalog # AAA16292-03
CAS Number:  50-23-7
Hydrocortisone 98%
Hydrocortisone 98%

Bottle, 1 g

CAS Number:  50-23-7
Price
$56.28
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A natural corticosteroid and anti-inflammatory compound

Purity: 98% CAS No.: 50-23-7 Molecular Formula: C21H30O5 Formula Weight: 362.47 Physical Form: Powder Appearance: White Odor: Odorless Melting Point: 213-215° Beilstein Reference: 1354819 Merck Reference: 14,4787 MDL No.: MFCD00011654

A natural corticosteroid and anti-inflammatory compound

  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Merck Index:14,4787
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:213…215 °C
  • Density:1.4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-020-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.