Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Basic fuchsin

Basic fuchsin

Bottle, 100 g
Catalog # AAA12952-22
Irritant
Health
Supplier #:  A12952-22
Basic fuchsin
Catalog # AAA12952-22
CAS Number:  632-99-5
Basic fuchsin
Basic fuchsin

Bottle, 100 g

CAS Number:  632-99-5
Price
$293.81
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Biological stain for distinguishing coli and aerogene bacteria

CAS No.: 632-99-5 Molecular Formula: C20H20ClN3 Formula Weight: 337.86 Melting Point: 268-270° dec. Solubility: Soluble in water Beilstein Reference: 4166684 Merck Reference: 14,5652 MDL No.: MFCD00012569

Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀ClN₃
  • MDL Number:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck Index:14,5652
  • Melting Pt:268…270 °C (dec.)
  • Density:1.22 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:211-189-6

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.