Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Adenosine 99%

Adenosine 99%

250g
Catalog # AAA10781-30
Supplier #:  A10781-30
Adenosine 99%
Catalog # AAA10781-30
CAS Number:  58-61-7
Adenosine 99%
Adenosine 99%

250g

CAS Number:  58-61-7
Price
$619.34
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 58-61-7 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Storage Temperature: 2°C to 8°C Physical Form: Powder Odor: Odorless Melting Point: 234-237° Beilstein Reference: 93029 Merck Reference: 14,153 Optical Rotation: -70° (c=2 in 5% NaOH) MDL No.: MFCD00005752

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Merck Index:14,153
  • Melting Pt:234…237 °C
  • Storage Temperature:Ambient
  • EINECS:200-389-9
Specifications

Specification Test Results

  • Optical rotation:
    -70° (c=2 in 5% NaOH)

Specifications

  • Size:
    250g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.