Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Silver nitrate standardised solution 0.1 N in aqueous solution

Silver nitrate standardised solution 0.1 N in aqueous solution

Drum, 20 L
Catalog # AA35601-M4
Corrosive
Environment
Supplier #:  035601-M4
Silver nitrate standardised solution 0.1 N in aqueous solution
Catalog # AA35601-M4
CAS Number:  7761-88-8
Silver nitrate standardised solution 0.1 N in aqueous solution
Silver nitrate standardised solution 0.1 N in aqueous solution

Drum, 20 L

CAS Number:  7761-88-8
Price
$2,984.14
Log in for availability
Each (20l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00003414

CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colorless Odor: Odorless UN No.: 3082 MDL No.: MFCD00003414

Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Drum
  • Size:
    20 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.