Orders LinkContactUsLinkComponent
Calcium oxide, REacton®, Reagent Grade
Bottle, 2 kg
  AA33299-A3
 :  033299-A3
Calcium oxide, REacton®, Reagent Grade
  AA33299-A3
 :  1305-78-8
Calcium oxide, REacton®, Reagent Grade
Bottle, 2 kg
 :  1305-78-8
$298.76
Each (2kg)
Manufacturing of steel, aluminum, magnesium, and glass and calcium salts, building and construction materials, lubricants, laboratory reagent
CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: Powder Density: 3.37 Refractive Index: 1.83 Sensitivity: Air and Moisture Sensitive Appearance: White Odor: Odorless Melting Point: 2572° Boiling Point: 2850° Solubility: Soluble in water with generation of heat, and forming Ca(OH)2, in acids, glycerol. Insoluble in alcohol Merck Reference: 14,1686 MDL No.: MFCD00010911
Manufacturing of steel, aluminum, magnesium, and glass and calcium salts, building and construction materials, lubricants, laboratory reagent
 :  Quicklime, Calcium oxide from marble, caustic, Lime
  •  :1305-78-8
  •  :CaO
  •  :MFCD00010911
  •  :56.08 g/mol
  •  :14,1686
  •  :2850 °C
  •  :2572 °C
  •  :3.37
  •  :Ambient
  •  :215-138-9
  •  :1910
  •  :8,III
  • Pack type:
    Bottle
  • Size:
    2 kg