Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium oxide, REacton®, Reagent Grade

Calcium oxide, REacton®, Reagent Grade

Bottle, 500 g
Catalog # AA33299-36
Corrosive
Irritant
Supplier #:  033299-36
Calcium oxide, REacton®, Reagent Grade
Catalog # AA33299-36
CAS Number:  1305-78-8
Calcium oxide, REacton®, Reagent Grade

Bottle, 500 g

CAS Number:  1305-78-8
Price
$154.96
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Manufacturing of steel, aluminum, magnesium, and glass and calcium salts, building and construction materials, lubricants, laboratory reagent

CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: Powder Density: 3.37 Refractive Index: 1.83 Sensitivity: Air and Moisture Sensitive Appearance: White Odor: Odorless Melting Point: 2572° Boiling Point: 2850° Solubility: Soluble in water with generation of heat, and forming Ca(OH)2, in acids, glycerol. Insoluble in alcohol Merck Reference: 14,1686 MDL No.: MFCD00010911

Manufacturing of steel, aluminum, magnesium, and glass and calcium salts, building and construction materials, lubricants, laboratory reagent

Synonyms:  Quicklime, Calcium oxide from marble, caustic, Lime
  • CAS Number:1305-78-8
  • Formula:CaO
  • MDL Number:MFCD00010911
  • Molecular Weight:56.08 g/mol
  • Merck Index:14,1686
  • Boiling Pt:2850 °C
  • Melting Pt:2572 °C
  • Density:3.37
  • Storage Temperature:Ambient
  • EINECS:215-138-9
  • UN:1910
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.