Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Selenium dioxide ≥99.4% (metals basis)

Selenium dioxide ≥99.4% (metals basis)

Bottle, 100 g
Catalog # AA12358-22
Toxic
Health
Environment
Supplier #:  012358-22
Selenium dioxide ≥99.4% (metals basis)
Catalog # AA12358-22
CAS Number:  7446-08-4
Selenium dioxide ≥99.4% (metals basis)

Bottle, 100 g

CAS Number:  7446-08-4
Price
$108.63
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

As an oxidizing agent, in manufacturing of other selenium compounds

Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odor: Odorless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

As an oxidizing agent, in manufacturing of other selenium compounds

  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:14,8434
  • Melting Pt:340…350 °C (subl.)
  • Density:3.95
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.