Orders LinkContactUsLinkComponent
ASTM Class 4 Precision Weight Sets
1 mg - 100 g, 21
  97021-882
 :  11124069
ASTM Class 4 Precision Weight Sets
  97021-882
ASTM Class 4 Precision Weight Sets
ASTM Class 4 Precision Weight Sets
1 mg - 100 g, 21
$2,570.00
Each
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
  • ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
  • Range:
    1 mg - 100 g
  • No. of weights:
    21