ASTM Class 4 Precision Weight Sets
10 mg - 50 g, 16
Catalog # 97021-876
Supplier: Mettler Toledo
: 11124080
Jump to:
Product Details & Documents
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
- 316 stainless steel
- Low magnetic permeability
- High corrosion resistance
- High gloss polish
- ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Specifications
- Range:10 mg - 50 g
- No. of weights:16