Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

NEFA Linearity Set Solution, Wako

NEFA-HR(2) Color B, 4×25 ml, +2…+10 °C
  97000-014
 :  991-34891
NEFA Linearity Set Solution, Wako
  97000-014
 :  
NEFA Linearity Set Solution, Wako
NEFA Linearity Set Solution, Wako

NEFA-HR(2) Color B, 4×25 ml, +2…+10 °C

 :  
 
$673.98
   
Each
 
   
 

The NEFA Linearity Set is a 2.5 mEq/L solution used to verify the measurable range of the NEFA HR(2) assay.

This product is only used with Wako’s NEFA HR(2) series of reagents.

 

 

 

  • Description:
    NEFA-HR(2) Color B
  • Size:
    4×25 ml
  • Storage temperature:
    +2…+10 °C
  • Cat. No.:
    97000-014
  • Linearity:
    0.01 - 4.00 mEq/L
  • Method:
    Enzymatic

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.