$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
DISINF HI-CON 256 NPD 30 GL
93002-002
:
Cani
DISINF HI-CON 256 NPD 30 GL
93002-002
:
Cani
:
CAN-H256-30
ghs_CAN-H256
VWR GHS Compliance Letter (US)