Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEDIUM TSA PENASE ST 100MM 32ML CS100
Catalog # 90006-622
Supplier:
BD
SDS
Certificates
Expand image
MEDIUM TSA PENASE ST 100MM 32ML CS100
Catalog # 90006-622
Supplier:
BD
Supplier Number:
215149
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_215149
Documentation