ContactUsLinkComponent
Protein-Free Blocking Buffer - Tbst (Tris-Buffered Saline, Ph 7.5 With 0.05% Tween*-20), 500 Ml
Protein-Free™ Blocking Buffers, G-Biosciences
Catalog # 89167-860
Supplier:  G-Biosciences
CAS Number:  
Protein-Free™ Blocking Buffers, G-Biosciences
Catalog # 89167-860
Supplier:  G-Biosciences
Supplier Number:  786-663
CAS Number:  
Specifications
  • Description:
    Protein-Free™Blocking Buffer-TBST
  • Cat. No.:
    89167-860
  • Supplier no.:
    786-663
Specifications
About this item
G-Biosciences' Protein-Free™ Blocking Buffer does not contain protein; it is a proprietary formation of non-protein agents that eliminates non-specific binding sites in ELISA, blotting, immunohistochemistry and other applications
G-Biosciences' Protein-Free™ Blocking Buffer eliminates any concern associated with regulatory compliance issues where use of animal source components are restricted. Furthermore, Protein-Free™ Blocking Buffer is compatible with antibodies and avidin/biotin based systems and results in high signal to background ratios.
G-Biosciences' Protein-Fre™ Blocking Buffer is offered in four convenient ready-to-use formats, with or without detergent: supplied either in TBS buffer (Tris-buffered saline at pH 7.5) or PBS buffer (phosphate-buffered saline at pH 7.5), as well as in separate formulations that contain G-Biosciences' detergent Tween®-20 for improving blocking efficiencies.
The absence of protein from the Protein-Free™ Blocking Buffer eliminates problems associated with traditional protein based blockers, such as cross-reactivity and interference from glycosylated proteins.
Ordering information: Accessories available.