Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Uridine

Uridine

50 g
Catalog # 89130-790
Supplier:   HiMedia
Supplier #:  TC181-50G
Uridine
Catalog # 89130-790
Supplier:  HiMedia
CAS Number:  58-96-8
Uridine

50 g

CAS Number:  58-96-8
Price
$195.00
Log in for availability
Each (50g)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
Specifications

Specification Test Results

  • Molecular Weight:
    244.2

Specifications

  • Size:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.