Orders LinkContactUsLinkComponent
Inoculation Loop, Globe Scientific
Inoculation Loop, Globe Scientific
Inoculation Loop, 1 uL and 10 uL, White, Sterile, 20/Peel Pack, 25 Packs/Case
Catalog # 60820-134
Supplier:   Globe Scientific
Supplier #:  2810
Inoculation Loop, Globe Scientific
Catalog # 60820-134
Supplier:  Globe Scientific
Inoculation Loop, Globe Scientific
Inoculation Loop, Globe Scientific
Inoculation Loop, 1 uL and 10 uL, White, Sterile, 20/Peel Pack, 25 Packs/Case
Price
$87.54
Log in for availability
Case of 500
Product Details & Documents
Loops offer the most convenient and safest way of streaking culture media
  • Polystyrene (PS)
  • Certificate of Calibration comes with each case of product
  • Gamma sterilized
  • Rigid feel for comfortable, stable, uniform streaking
  • One side is a 10 µl loop and the reverse side is a 1 µl loop
Loops are sterilized by gamma radiation.
Molded from a special medical grade polystyrene, they offer a strong, rigid feel to provide comfortable, stable, and uniform streaking.
Delivery information: Supplied with calibration certificate.
Documents
Specifications
  • Sterility:
    Sterile
  • Color:
    White
  • Description:
    Inoculation Loop, 1 uL and 10 uL
  • Packaging:
    20/Peel Pack, 25 Packs/Case