Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MOUNTING MEDIUM CLEAR PK=5UNIT
Catalog # 40176-126
Supplier:
Fulton Metallurgical Products
SDS
Expand image
MOUNTING MEDIUM CLEAR PK=5UNIT
Catalog # 40176-126
Supplier:
Fulton Metallurgical Products
Supplier Number:
5 UNIT PACK
SDS
Related Information
Documentation
SDS
ghs_5 UNIT PACK.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation