Supor® Membrane Disc Filters
Product Details & Documents
Supor® hydrophilic polyethersulfone membranes offer higher fluid flow rates, lower protein binding, lower extractables, and greater strength than other cellulosic (acetate, nitrate) membranes.
- Hydrophilic
- Higher flow rate
- Stronger than cellulose
Provide some of the highest water flow rates and fluid filtration capacities of any membrane filter. These membranes are non sterile, except for 28147-979, which is gamma-irradiated.
The 142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number.
Several of these membranes are offered as recommended replacements for the discontinued HT Tuffyrn membranes.
Documents
Certificates
Search by Lot NumberPall Laboratory Chemical Compatibility Chart Pall Laboratory Microbiology QC Products Catalog PALL SolVac® Filter Holder
Product Literature
Pall Laboratory Microbiology QC products
Promotion Material
Specifications
- Ø:142 mm
- Pore size:0.45 µm
- Sterility:Non sterile
- Description:Plain, Tabbed
- Gamma-irradiated:No
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



