Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Eppendorf® Safe-Lock Tubes, 0.5 ml - Microtubes

Eppendorf® Safe-Lock Tubes, 0.5 ml - Microtubes

Amber, 30000×g, 500 tubes (1 bag×500 tubes)
Catalog # 20901-510
Product available on GSA Advantage®
Supplier:   Eppendorf
Supplier #:  022363638
Eppendorf® Safe-Lock Tubes, 0.5 ml - Microtubes
Catalog # 20901-510
Supplier:  Eppendorf
Product available on GSA Advantage®
Eppendorf® Safe-Lock Tubes, 0.5 ml - Microtubes
Eppendorf® Safe-Lock Tubes, 0.5 ml - Microtubes

Amber, 30000×g, 500 tubes (1 bag×500 tubes)

Price
$128.00
Log in for availability
Case of 500
Product Details & Documents

Eppendorf® Safe-Lock tubes 0.5 ml - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

  • Hinged Safe-Lock lid prevents unintended opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss in the event of tube breakage, and provides enhanced safety when handling hazardous samples
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures
  • Precise lid sealing minimizes evaporation rates during long-term storage
  • Large frosted lid and side surface facilitate easy labeling
  • Ensured functionality in a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for effective sterilization
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur® quality, ensuring tested purity

Eppendorf® Safe-Lock 0.5 ml microtubes epitomize advanced sample storage, featuring a patented Safe-Lock design for unparalleled security. Tailored for PCR applications, these microtubes provide a space-efficient 0.5 ml capacity, ideal for labs with limited storage. Their compatibility with various centrifuge models ensures seamless integration into existing workflows. Robust construction and user-friendly attributes, such as clear graduations, further enhance reliability and ease of handling. Elevate laboratory practices with Eppendorf® Safe-Lock tubes, which set a new standard for sample integrity and operational efficiency.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Cap type: Safe-Lock
Material: PP
Volume: 0.5 ml

 : Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Specifications
  • Color:
    Amber
  • Packaging:
    500 tubes (1 bag×500 tubes)
  • Max. rcf:
    30000×g
  • Cat. No.:
    20901-510
  • Temperature Range:
    −86…+100 °C
  • Autoclavable:
    When open 121 °C, 20 mins