Orders LinkContactUsLinkComponent
Simultaneous Test Sets, Drager Safety
Simultaneous Test Sets, Drager Safety
Simultaneous test set III, for organic vapors (ketone, aromates, alcohol, aliphatic hydrocarbons, chlorinated hydrocarbons), 120 s
Catalog # 13453-128
Supplier:   Draeger
Supplier #:  8101770
Simultaneous Test Sets, Drager Safety
Catalog # 13453-128
Supplier:  Draeger
Simultaneous Test Sets, Drager Safety
Simultaneous Test Sets, Drager Safety
Simultaneous test set III, for organic vapors (ketone, aromates, alcohol, aliphatic hydrocarbons, chlorinated hydrocarbons), 120 s
Price
$107.68
Log in for availability
Pack of 10
Minimum order quantity is 1
Product Details & Documents
Risks and potential dangers at a glance: The SimultanTest Sets allow the measurement of five different gases at the same time.
In the SimultanTest Set, five Dräger-Tubes are arranged next to each other in a stable rubber sleeve. The air to be monitored is simultaneously drawn in through the tubes by a Dräger-Tube pump. Quick determinations are made about the presence of dangerous substances at TLV levels.
The device delivers fast and reliable results for different combinations of organic and inorganic gases.
Documents
Specifications
  • Description:
    Simultaneous test set III, for organic vapors (ketone, aromates, alcohol, aliphatic hydrocarbons, chlorinated hydrocarbons)
  • Response time:
    120 s
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Adapter for Simultaneous test sets, for connection to a bellows pump
    Adapter for Simultaneous test sets, for connection to a bellows pump
    34330-044
    Log in for availability
    Each - $116.26
Product Information
  • Catalog #
    Availability
    Price
  • Extension Hose
    Extension Hose
    13450-858
    Log in for availability
    Each - $319.86