Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REVISED USP 467 CAL MIX EA4
Catalog # ULUSPM-467C
Supplier:
Ultra Scientific
SDS
Expand image
REVISED USP 467 CAL MIX EA4
Catalog # ULUSPM-467C
Supplier:
Ultra Scientific
Supplier Number:
USPM-467C
SDS
Related Information
Documentation
SDS
ghs_USPM-467C
Documentation