$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
AVIATION GAS SOLUTION
ULRGO-662-1
:
Ultra Scientific
AVIATION GAS SOLUTION
ULRGO-662-1
:
Ultra Scientific
:
RGO-662-1
ghs_RGO-662-1.pdf
VWR GHS Compliance Letter (US)