Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
METHOD 529 CAL STANDARD PK4
Catalog # ULNAIM-529A
Supplier:
Ultra Scientific
SDS
Expand image
METHOD 529 CAL STANDARD PK4
Catalog # ULNAIM-529A
Supplier:
Ultra Scientific
Supplier Number:
NAIM-529A
SDS
Related Information
Documentation
SDS
ghs_NAIM-529A
Documentation