$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
F003 SOLVENT LIST MIXTURE
ULFLM-003-1
:
Ultra Scientific
F003 SOLVENT LIST MIXTURE
ULFLM-003-1
:
Ultra Scientific
:
FLM-003-1
ghs_FLM-003-1.pdf
VWR GHS Compliance Letter (US)