Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Vanillin ≥98.0%

25 g
  TCH0264-025G
 
 :  H0264-25G
Vanillin ≥98.0%
  TCH0264-025G
 :  121-33-5
Vanillin ≥98.0%

25 g

 :  121-33-5
 
$34.00
   
Each (25g)
 
   
 

CAS Number: 121-33-5
MDL Number: MFCD00006942
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 284
Melting point (°C): 83
Flash Point (°C): 158

Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Others (Phytochemicals)
Other Product Names / Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

 :  4-Hydroxy-3-methoxybenzaldehyde
  •  :121-33-5
  •  :C₈H₈O₃
  •  :MFCD00006942
  •  :152.15 g/mol
  •  :13,09998
  •  :284 °C
  •  :83 °C
  •  :1.056 g/cm³ (20 °C)
  •  :158 °C
  •  :Ambient
  •  :204-465-2
 

 

  • Purity/Analysis Method:
    >98.0% (GC,T)

 

  • Size:
    25 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.