Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
2-Phenoxyethanol

2-Phenoxyethanol

4 L, Glass bottle, amber, narrow mouth
Catalog # JTT319-9
Irritant
Supplier #:  T319-09
2-Phenoxyethanol
Catalog # JTT319-9
CAS Number:  122-99-6
MOC:  Non-RegulatedCertificates
2-Phenoxyethanol
2-Phenoxyethanol

4 L, Glass bottle, amber, narrow mouth

CAS Number:  122-99-6
MOC:  Non-Regulated
Price
$289.15
Log in for availability
Each (4l) Case of 4 (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Merck Index:12,07410
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:13 °C
  • Density:1.1094 g/cm³ (20 °C)
  •  :Management of Change (MOC) category = N
  • Flash Pt:121 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:204-589-7

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Identification (by IR):
    Passes Test

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    4 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.