2-Methoxypropene 95% stabilised
Product Details & Documents
Fieser: 2,230 5,360 12,317
Purity: 95% CAS No.: 116-11-0 Molecular Formula: C4H8O Formula Weight: 72.11 Storage Temperature: 2°C to 8°C Flash Point: -29°(-20°F) Refractive Index: 1.3835 Odor: Ether-like Boiling Point: 35-36° Beilstein Reference: 1734635 UN No.: 1993 MDL No.: MFCD00014929
- CAS Number:116-11-0
- Formula:C₄H₈O
- MDL Number:MFCD00014929
- Molecular Weight:72.11 g/mol
- Boiling Pt:35…36 °C
- Density:0.758
- Flash Pt:–29 °C (–20 °F)
- Flash Pt:-29 °C(-20 °F)
- Storage Temperature:Ambient
- EINECS:204-125-3
- UN:1993
- ADR:3,III
- :Stabilised with potassium carbonate ∼0.5%
- ADR:"3",III
Specifications
Specifications
- Size:50 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



