Chlorophenol red
Bottle, 5 g
Supplier #: B21623-06
CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877
: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- :4430-20-0
- :C₁₉H₁₂Cl₂O₅S
- :MFCD00005877
- :423.27 g/mol
- :∼261 °C
- :Ambient
- :224-619-2
Documents
- Pack type:Bottle
- Size:5 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



