Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Valeric acid 99%

Bottle, 500 ml
Catalog # AAA16238-AP
Corrosive
 :  A16238-AP
Valeric acid 99%
Catalog # AAA16238-AP
CAS Number:  109-52-4
Valeric acid 99%

Bottle, 500 ml

CAS Number:  109-52-4
Price
$61.46
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 109-52-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 88°(190°F) Refractive Index: 1.4080 Odor: Unpleasant Melting Point: -34° Boiling Point: 184-186° Beilstein Reference: 969454 Merck Reference: 14,9904 UN No.: 3265 MDL No.: MFCD00004413

Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Merck Index:14,9904
  • Boiling Pt:184…186 °C
  • Melting Pt:–34 °C
  • Density:0.938
  • Flash Pt:88 °C (190 °F)
  • Storage Temperature:Ambient
  • EINECS:203-677-2
  • UN:3265
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.