Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bismuth(III) hydroxide, tech.

Bismuth(III) hydroxide, tech.

250 g
Catalog # AA40397-30
Irritant
Supplier #:  040397-30
Bismuth(III) hydroxide, tech.
Catalog # AA40397-30
CAS Number:  10361-43-0
Bismuth(III) hydroxide, tech.

250 g

CAS Number:  10361-43-0
Price
$119.84
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Applications: Hydrolysis of ribonucleic acid, absorbent
Insoluble in water. Soluble in acids

CAS No.: 10361-43-0 Molecular Formula: Bi(OH)3 Formula Weight: 260.00 Physical Form: Amorphous Powder Odor: Odorless Solubility: Insoluble in water. Soluble in acids MDL No.: MFCD00046188

Hydrolysis of ribonucleic acid, absorbent

Synonyms:  Bismuth hydroxide
  • CAS Number:10361-43-0
  • Formula:H₃BiO₃
  • MDL Number:MFCD00046188
  • Molecular Weight:260
  • Storage Temperature:Ambient
  • EINECS:233-790-2

Documents

Specifications

Specifications

  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.