D-(-)-Mannitol ≥99.0% ACS
Product Details & Documents
MDL: MFCD00064287 Beilstein Registry No.: 1721898 Soluble in water, alcohol, pyridine, aniline. Insoluble in ether
CAS No.: 69-65-8 Molecular Formula: HOCH2(CHOH)4CH2OH Formula Weight: 182.17 Physical Form: Crystalline Density: 1.489 Appearance: White Odor: Odorless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Solubility: Soluble in water, alcohol, pyridine, aniline. Insoluble in ether Beilstein Reference: 1721898 Merck Reference: 14,5745 MDL No.: MFCD00064287
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:14,5745
- Boiling Pt:290…295 °C (3.5 mmHg)
- Melting Pt:165…167 °C
- Density:1.489
- Storage Temperature:Ambient
- EINECS:200-711-8
Specifications
Specification Test Results
- Insoluble matter:0.01%
- Residue after ignition:0.01%
- Titratable acid:0.0008meq/g
- Reducing sugars:Pass test
- Heavy Metals (as Pb):5 ppm
- Specific rotation at 25°:+23.3° to +24.3°
Specifications
- Pack type:Bottle
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



