VWR® Peroxidase
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
HRP catalyzes the decomposition of hydrogen peroxide in solution causing the oxidation of a number of substrates.
Used extensively as a label for immunohistological assays and ELISA. Its exceptional stability in powdered and solution form enhances its desirability as an enzyme conjugate.
Documents
Specifications
Specifications
- Pack type:Bottle, amber
- Size:10000 units
- Storage Conditions:Freezer
- Cat. No.:97061-454
- Tested Applications:for biochemistry
- MDL No.:MFCD00071339
- Enzyme Name:Peroxidase
- CAS No.:9003-99-0
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



