Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Electrode Fill Solutions

Electrode Fill Solutions

1M KNO₃, Bottle, 25 ml, Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide
Catalog # 97001-718
Supplier #:  51343182
Electrode Fill Solutions
Catalog # 97001-718
CAS Number:  
Electrode Fill Solutions
Electrode Fill Solutions

1M KNO₃, Bottle, 25 ml, Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide

CAS Number:  
Price
$26.80
Log in for availability
Each (25ml)
Product Details & Documents

FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.

Delivery information: Supplied in 25 ml (0.9 oz.) bottles.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    1M KNO₃
  • Ion:
    Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide
  • Volume:
    25 ml

Specifications