Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DISINF HI-CON 256 NPD 5 GL
Catalog # 93002-004
Supplier:
Cani
SDS
Expand image
DISINF HI-CON 256 NPD 5 GL
Catalog # 93002-004
Supplier:
Cani
Supplier Number:
CAN-H256-5
SDS
Related Information
Documentation
SDS
ghs_CAN-H256
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation