Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Difco™ YPD Medium, BD Life Sciences

Difco™ YPD Medium, BD Life Sciences

YPD Agar, 500 g
Catalog # 90003-280
Supplier:   BD
Supplier #:  242720
Difco™ YPD Medium, BD Life Sciences
Catalog # 90003-280
Supplier:  BD
CAS Number:  
Difco™ YPD Medium, BD Life Sciences
Difco™ YPD Medium, BD Life Sciences

YPD Agar, 500 g

CAS Number:  
Price
$241.00
Log in for availability
Each (500g)
Product Details & Documents

For propagation and maintenance of yeasts for molecular biology applications.

  • Dehydrated culture media
Specifications

Specifications

  • Description:
    YPD Agar
  • Size:
    500 g

Specifications