Orders LinkContactUsLinkComponent
Histosette® I Processing and Embedding Cassettes, Simport
Histosette® I Processing and Embedding Cassettes, Simport
Biopsy Cassettes, Aqua
Catalog # 87002-432
Supplier:   Simport Scientific
Supplier #:  M491-12
Histosette® I Processing and Embedding Cassettes, Simport
Catalog # 87002-432
Supplier:  Simport Scientific
Histosette® I Processing and Embedding Cassettes, Simport
Histosette® I Processing and Embedding Cassettes, Simport
Biopsy Cassettes, Aqua
Price
$457.43
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable plastic cassettes with lids are made of acetal polymer and hold tissue or biopsy specimens during the embedding process and for storage.
They keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The lid can be easily snapped apart and locked into the base of the cassette. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 30° angle. The biopsy cassettes feature 1mm square openings.
Ordering information: Cassettes are available in 11 non cytotoxic, non metallic colors, and three fluorescent colors. Each case contains three dispenser boxes of 500 cassettes.
Documents
Specifications
  • Color:
    Aqua
  • Description:
    Biopsy Cassettes