Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Smartwall G3™ Productivity System, Metro™

Metroseal green epoxy storage basket, Super Erecta shelving, Metroseal 3, 5", 13.375", 7"
Catalog # 83009-462
Supplier:   Metro
 :  H209K3
Smartwall G3™ Productivity System, Metro™
Catalog # 83009-462
Supplier:  Metro
Smartwall G3™ Productivity System, Metro™
Smartwall G3™ Productivity System, Metro™

Metroseal green epoxy storage basket, Super Erecta shelving, Metroseal 3, 5", 13.375", 7"

Price
$68.50
Log in for availability
Each
 
Product Details & Documents

Smartwall G3™ Productivity System provides for organized, efficient wall space.

  • Shelves, shelf supports, grids and accessories can easily be removed for thorough cleaning of walls
  • Wall tracks and uprights can easily be wiped down

Choose from Super Erecta Brite for dry environments or Metroseal 3 epoxy for wet or damp environments. Metroseal 3 features built in Microban® antimicrobial product protection to inhibit the growth of bacteria, mold, mildew and fungus that cause odor and product degradation.

Wall mounted tracks are the foundation for attaching productivity stations with grids, accessories and storage shelves. Smartwall G3™ system is ideal for prep areas, janitorial supply areas, above carts in staging areas or storage rooms and above sinks, work tables, casework, equipment and bulk floor storage.

 : For a complete list of Smartwall G3™ components and accessories, contact your Avantor representative. Metroseal 3 components have a 12 year warranty against rust and corrosion. Other components have manufacturers standard one year warranty.

 

Specifications
  • Finish:
    Metroseal 3
  • Width:
    5"
  • Description:
    Metroseal green epoxy storage basket
  • Height:
    7"
  • Length:
    13.375"
  • For use with:
    Super Erecta shelving
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.