Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tryptone Base Materials

Tryptone Base Materials

Tryptone, Glass bottle for solids, 500 g
Catalog # 71003-498
Supplier:   G-Biosciences
Supplier #:  RC-110
Tryptone Base Materials
Catalog # 71003-498
Supplier:  G-Biosciences
CAS Number:  
Tryptone Base Materials

Tryptone, Glass bottle for solids, 500 g

CAS Number:  
Price
$214.82
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for the growth of E. coli and other microorganisms, as it provides a source of amino acids.

  • Solubility: Water

  • Appearance: Cream to beige free flowing powder
  • CAS: 91079-40-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Description:
    Tryptone
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.