Xcelite® Precision Electronic Screwdriver Sets, Apex
TORX® Set
Catalog # 37699-592
Supplier: Cooper Tools
Supplier #: XPTX600
Jump to:
Product Details & Documents
For protection of ESD-sensitive items
The chrome-molybdenum-vanadium steel blades provide superior strength. Screwdrivers feature fast turning swivel caps with cavity to provide optimum control for precise centering, black oxide tips for longer wear life, no-roll handle design, and a storage case.
Set 37699-570 includes one each of No. 0 and No. 00 point Phillips® screwdrivers and slotted tip screwdrivers of the following sizes: 1.5, 2, 2.5, and 3 mm. TORX® set 37699-592 includes one each of No. 5 point and No. 6 point standard screwdrivers and four tamper-proof standard screwdrivers: No. 7, No. 8, No. 10, and No. 15 point.
Specifications
- Description:TORX® Set