Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TITRANT KF STABLE BOTTLE 5MG
Catalog # 21432-344
Supplier:
GFS Chemicals
SDS
Expand image
TITRANT KF STABLE BOTTLE 5MG
Catalog # 21432-344
Supplier:
GFS Chemicals
Supplier Number:
99661
SDS
Related Information
Documentation
SDS
ghs_1604.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation