$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
SCREEN+KEYBRD CLNR 1QT CS12
13502-348
:
ITW Chemtronics
SCREEN+KEYBRD CLNR 1QT CS12
13502-348
:
ITW Chemtronics
:
1743-QT
ghs_1743.pdf
VWR GHS Compliance Letter (US)