$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
SIMAZINE SOLUTION
ULPP-530A-1
:
Ultra Scientific
SIMAZINE SOLUTION
ULPP-530A-1
:
Ultra Scientific
:
PP-530A-1
ghs_PP-530A-1.pdf
VWR GHS Compliance Letter (US)