Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Nuclear Fast Red

10 g
Catalog # TCN0184-010G
Supplier:   TCI America
 :  N0184-10G
Nuclear Fast Red
Catalog # TCN0184-010G
Supplier:  TCI America
CAS Number:  6409-77-4
Nuclear Fast Red

10 g

CAS Number:  6409-77-4
Price
$302.00
Log in for availability
Each (10g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 6409-77-4
MDL Number: MFCD00078493
Molecular Formula: C14H9NO7S
Molecular Weight: 357.27
Form: Crystal
Color: Deep Yellow Red

Other Product Names / Synonyms: Kernechtrot, NFR

Synonyms:  CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
  • CAS Number:6409-77-4
  • Formula:C₁₄H₈NNaO₇S
  • MDL Number:MFCD00078493
  • Molecular Weight:357.28 g/mol
  • Merck Index:13,06512
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:229-088-0
Specifications

Specifications

  • Size:
    10 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.